Assay Details
Target Gene Details
Entrez Gene ID: | 547253 |
Gene Name: | poly (ADP-ribose) polymerase family, member 14 |
Gene Aliases: |
1600029O10Rik, ARTD8, BC021340, CoaSt6, mKIAA1268 |
Location: |
Chr.16:35832859-35871514 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp14 | NM_001039530.3 | NP_001034619.2 | ||
| AK089685.1 | BAC40943.1 | |||
| AK220263.1 | BAD90188.1 | |||
| BC021340.1 | AAH21340.1 | |||
| BC138874.1 | ||||
| DQ372930.1 | ABD16173.1 |

Genomic Map