Assay Details
Target Gene Details
Entrez Gene ID: | 547253 |
Gene Name: | poly (ADP-ribose) polymerase family, member 14 |
Gene Aliases: |
1600029O10Rik, ARTD8, BC021340, CoaSt6, mKIAA1268 |
Location: |
Chr.16:35832859-35871514 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp14 | NM_001039530.3 | 8 | 3495 | NP_001034619.2 |
| AK089685.1 | 9 | 3586 | BAC40943.1 | |
| AK220263.1 | 3 | 1087 | BAD90188.1 | |
| BC138874.1 | 8 | 3610 | ||
| DQ372930.1 | 8 | 3659 | ABD16173.1 |

Genomic Map