Assay Details
Target Gene Details
Entrez Gene ID: | 56370 |
Gene Name: | transgelin 3 |
Gene Aliases: |
2700038H05Rik, 2900005O10Rik, AI426007, Np25 |
Location: |
Chr.16:45711230-45724531 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tagln3 | NM_019754.3 | NP_062728.1 | ||
| AB031291.1 | BAA83499.1 | |||
| AK012333.2 | BAC25365.1 | |||
| AK013484.1 | BAB28876.1 | |||
| BC043027.1 | AAH43027.1 | |||
| BC055338.1 | AAH55338.1 |

Genomic Map