Assay Details
Target Gene Details
Entrez Gene ID: | 80285 |
Gene Name: | poly (ADP-ribose) polymerase family, member 9 |
Gene Aliases: |
ARTD9, AW214463, BC003281, Bagl, Bal, PARP-9 |
Location: |
Chr.16:35938470-35972621 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Parp9 | NM_030253.3 | NP_084529.1 | ||
| AK037903.1 | BAC29897.1 | |||
| AK050032.1 | BAC34040.1 | |||
| BC003281.1 | AAH03281.1 | |||
| BC070466.1 | AAH70466.1 |
More Information
Set Membership: |
|

Genomic Map