Assay Details
Target Gene Details
Entrez Gene ID: | 76302 |
Gene Name: | PEST proteolytic signal containing nuclear protein |
Gene Aliases: |
1110018D06Rik, AI647035 |
Location: |
Chr.16:56015506-56030177 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcnp | NM_001024622.2 | NP_001019793.1 | ||
| AK145195.1 | BAE26287.1 | |||
| AK150595.1 | BAE29688.1 | |||
| AK152279.1 | BAE31093.1 | |||
| AK159201.1 | BAE34895.1 | |||
| AK166071.1 | BAE38553.1 | |||
| AK168723.1 | ||||
| AK172325.1 | ||||
| BC061235.1 | AAH61235.1 | |||
| BC106192.1 | AAI06193.1 | |||
| BC132588.1 | ||||
| BC132590.1 | ||||
| BY287179.1 |

Genomic Map