Assay Details
Target Gene Details
Entrez Gene ID: | 17880 |
Gene Name: | myosin, heavy polypeptide 11, smooth muscle |
Gene Aliases: |
AV071570, SM1, SM2, smMHC |
Location: |
Chr.16:14194527-14291408 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myh11 | NM_001161775.1 | NP_001155247.1 | ||
| NM_013607.2 | NP_038635.2 | |||
| AK137226.1 | ||||
| AK137716.1 | ||||
| AK173045.1 | BAD32323.1 | |||
| BC026142.1 | AAH26142.1 | |||
| D85923.1 | BAA19690.1 | |||
| D85924.1 | BAA19691.1 |

Genomic Map