Assay Details
Target Gene Details
Entrez Gene ID: | 52633 |
Gene Name: | nitrilase family, member 2 |
Gene Aliases: |
1190017B19Rik, D16Ertd502e |
Location: |
Chr.16:57156665-57167332 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nit2 | NM_023175.1 | NP_075664.1 | ||
| AF284573.1 | AAF87102.1 | |||
| AK003604.1 | BAB22884.1 | |||
| AK004533.1 | ||||
| AK004535.1 | BAB23354.1 | |||
| BC020153.1 | AAH20153.1 |
More Information
Set Membership: |
|

Genomic Map