Assay Details
Target Gene Details
Entrez Gene ID: | 18951 |
Gene Name: | septin 5 |
Gene Aliases: |
Cdcrel-1, Cdcrel1, Pnutl1 |
Location: |
Chr.16:18621811-18629938 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sept5 | NM_213614.2 | NP_998779.2 | ||
| AF033350.1 | AAC83974.1 | |||
| AK134700.1 | BAE22248.1 | |||
| BC059848.1 | AAH59848.1 | |||
| BC094347.1 | AAH94347.1 | |||
| BC141073.1 | ||||
| BC145331.1 | ||||
| BC145332.1 | ||||
| CA945297.1 |
More Information
Set Membership: |
|

Genomic Map