Assay Details
Target Gene Details
Entrez Gene ID: | 16180 |
Gene Name: | interleukin 1 receptor accessory protein |
Gene Aliases: |
6430709H04Rik, AI255955, AV239853, IL-1RAcP |
Location: |
Chr.16:26581688-26730128 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1rap | NM_001159317.1 | NP_001152789.1 | ||
| NM_001159318.1 | NP_001152790.1 | |||
| NM_008364.2 | NP_032390.1 | |||
| AK033972.1 | ||||
| AK034269.1 | ||||
| AK045686.1 | BAC32457.1 | |||
| AK083209.1 | BAE43382.1 | |||
| AK136782.1 | BAE23128.1 | |||
| X85999.1 | CAA59991.1 |
More Information
Set Membership: |
|

Genomic Map