Assay Details
Target Gene Details
Entrez Gene ID: | 13026 |
Gene Name: | phosphate cytidylyltransferase 1, choline, alpha isoform |
Gene Aliases: |
CTalpha, Cctalpha, Ctpct, Cttalpha |
Location: |
Chr.16:32430921-32475065 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcyt1a | NM_001163159.1 | NP_001156631.1 | ||
| NM_009981.4 | NP_034111.1 | |||
| AK044585.1 | ||||
| AK076050.1 | BAC36148.1 | |||
| BB642590.1 | ||||
| L28956.1 | AAA53526.1 | |||
| Z12302.1 | CAA78172.1 |
More Information
Set Membership: |
|

Genomic Map