Assay Details
Target Gene Details
Entrez Gene ID: | 66061 |
Gene Name: | Tctex1 domain containing 2 |
Gene Aliases: |
0610012D17Rik, 2410011K15Rik |
Location: |
Chr.16:32419686-32429099 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tctex1d2 | NM_025329.3 | NP_079605.1 | ||
| AK002589.1 | BAB22210.1 | |||
| AK004191.1 | BAB23214.1 | |||
| AK007178.1 | BAB24886.1 | |||
| AK010027.1 | BAB26652.1 | |||
| AK010464.1 | BAB26959.1 | |||
| BC059716.1 | AAH59716.1 |
More Information
Set Membership: |
|

Genomic Map