Assay Details
Target Gene Details
Entrez Gene ID: | 12915 |
Gene Name: | activating transcription factor 6 beta |
Gene Aliases: |
AA617266, ATF6beta, Creb-rp, Crebl1 |
Location: |
Chr.17:34647146-34655074 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atf6b | NM_017406.4 | NP_059102.2 | ||
| AK090378.1 | ||||
| AK143765.1 | BAE25530.1 | |||
| AK146671.1 | BAE27348.1 | |||
| AK159169.1 | BAE34870.1 | |||
| AK170027.1 | ||||
| BC013534.1 | AAH13534.1 | |||
| BC052635.1 | AAH52635.1 |
More Information
Set Membership: |
|

Genomic Map