Assay Details
Target Gene Details
Entrez Gene ID: | 53895 |
Gene Name: | caseinolytic mitochondrial matrix peptidase proteolytic subunit |
Gene Aliases: |
AU019820, D17Wsu160e |
Location: |
Chr.17:56990264-56996371 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clpp | NM_017393.2 | NP_059089.1 | ||
| AJ005253.1 | CAA06443.1 | |||
| AK002238.1 | BAC24984.1 | |||
| AK004024.1 | BAB23132.1 | |||
| AK145765.1 | BAE26636.1 | |||
| AK147719.1 | BAE28093.1 | |||
| AK168053.1 | BAE40033.1 | |||
| AK179220.1 | ||||
| BC001998.1 | AAH01998.1 |
More Information
Set Membership: |
|

Genomic Map