Assay Details
Target Gene Details
Entrez Gene ID: | 67544 |
Gene Name: | family with sequence similarity 120, member B |
Gene Aliases: |
4932442K08Rik, AW049283, AW413338, CCPG, Pgcc1, mKIAA1838 |
Location: |
Chr.17:15396157-15433581 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam120b | NM_024203.3 | NP_077165.2 | ||
| NR_033586.1 | ||||
| AK005957.1 | BAB24339.1 | |||
| AK016568.1 | ||||
| AK132211.1 | BAE21035.1 | |||
| BC003200.1 | AAH03200.1 | |||
| BC085163.1 | AAH85163.1 | |||
| DQ873694.1 |

Genomic Map