Assay Details
Target Gene Details
Entrez Gene ID: | 11596 |
Gene Name: | advanced glycosylation end product-specific receptor |
Gene Aliases: |
RAGE |
Location: |
Chr.17:34597410-34600937 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ager | NM_001271422.1 | 3 | 295 | NP_001258351.1 |
| NM_001271423.1 | 3 | 268 | NP_001258352.1 | |
| NM_001271424.1 | NP_001258353.1 | |||
| NM_007425.3 | 3 | 295 | NP_031451.2 | |
| NR_073173.1 | 3 | 295 | ||
| NR_073175.1 | 2 | 490 | ||
| NR_073176.1 | 2 | 490 | ||
| AB207883.1 | 3 | 255 | BAE72665.1 | |
| AK004802.1 | 3 | 283 | ||
| BC061182.1 | 3 | 283 | AAH61182.1 | |
| EU520325.1 | 3 | 251 | ||
| EU570240.1 | 3 | 251 | ||
| EU570241.1 | 2 | 446 | ||
| EU570242.1 | 3 | 251 | ||
| EU570243.1 | 3 | 251 | ||
| EU570244.1 | 2 | 446 | ||
| EU570245.1 | 2 | 446 | ||
| EU570246.1 | 3 | 251 | ||
| EU570247.1 | 2 | 446 | ||
| EU906857.1 | ||||
| EU906858.1 | 3 | 251 | ||
| EU906859.1 | 3 | 251 | ||
| EU906860.1 | ||||
| EU906861.1 | ||||
| EU906862.1 | 3 | 251 | ||
| EU906863.1 | 3 | 224 | ||
| EU906864.1 | 2 | 446 | ||
| GU164727.1 | ||||
| GU164728.1 | 3 | 270 | ||
| GU164729.1 | 3 | 270 | ||
| GU164730.1 | 3 | 270 | ||
| GU164731.1 | 3 | 270 | ||
| GU164732.1 | 3 | 270 | ||
| GU164733.1 | ||||
| GU164734.1 | 3 | 271 | ||
| GU164735.1 | ||||
| GU164736.1 | ||||
| GU164737.1 | ||||
| GU164738.1 | ||||
| GU164739.1 | ||||
| KC692918.1 | 3 | 251 | ||
| L33412.1 | 3 | 256 | AAA40040.1 |
More Information
Set Membership: |
|

Genomic Map