Assay Details
Target Gene Details
Entrez Gene ID: | 320159 |
Gene Name: | family with sequence similarity 179, member A |
Gene Aliases: |
4632412N22Rik |
Location: |
Chr.17:71673157-71729669 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam179a | NM_177087.4 | 14 | 2237 | NP_796061.2 |
| AK076284.1 | 13 | 1892 | ||
| AK158647.1 | 14 | 2022 | BAE34597.1 |
More Information
Set Membership: |
|

Genomic Map