Assay Details
Target Gene Details
Entrez Gene ID: | 66237 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit G2 |
Gene Aliases: |
1500002D01Rik, Atp6g2, NG38, VAG2 |
Location: |
Chr.17:35235166-35238767 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v1g2 | NM_023179.3 | 1 | 162 | NP_075668.1 |
| AB076405.1 | 1 | 216 | BAC10288.1 | |
| AK005109.1 | 1 | 43 | ||
| AK011945.1 | BAB27931.1 | |||
| BC020190.1 | 1 | 158 | AAH20190.1 | |
| BC062380.1 | 1 | 150 | AAH62380.1 | |
| BU558667.1 | 1 | 165 |
More Information
Set Membership: |
|

Genomic Map