Assay Details
Target Gene Details
Entrez Gene ID: | 22268 |
Gene Name: | uroplakin 1B |
Gene Aliases: |
AI413235, Tspan20, UPIb, Upk1 |
Location: |
Chr.16:38773184-38800345 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Upk1b | NM_178924.4 | 6 | 686 | NP_849255.2 |
| AF274865.1 | 5 | 621 | AAM97593.1 | |
| AK053004.1 | 6 | 687 | BAC35237.1 | |
| AK083101.1 | 6 | 686 | BAC38760.1 | |
| AK086926.1 | 6 | 685 | BAC39763.1 | |
| BC046604.1 | 6 | 664 | AAH46604.1 |

Genomic Map