Assay Details
Target Gene Details
Entrez Gene ID: | 22268 |
Gene Name: | uroplakin 1B |
Gene Aliases: |
AI413235, Tspan20, UPIb, Upk1 |
Location: |
Chr.16:38773184-38800345 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Upk1b | NM_178924.4 | NP_849255.2 | ||
| AF274865.1 | AAM97593.1 | |||
| AK053004.1 | BAC35237.1 | |||
| AK083101.1 | BAC38760.1 | |||
| AK086926.1 | BAC39763.1 | |||
| BC046604.1 | AAH46604.1 |

Genomic Map