Assay Details
Target Gene Details
Entrez Gene ID: | 68617 |
Gene Name: | microtubule crosslinking factor 1 |
Gene Aliases: |
1110012J17Rik, Ccdc165, D130071O13, Kiaa0802, Soga2, mKIAA0802, t8219b25 |
Location: |
Chr.17:66336980-66449808 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mtcl1 | NM_001114098.1 | NP_001107570.1 | ||
| NM_172963.4 | NP_766551.3 | |||
| AK147205.1 | BAE27762.1 | |||
| AK147631.1 | ||||
| AK147691.1 |

Genomic Map