Assay Details
Target Gene Details
Entrez Gene ID: | 57261 |
Gene Name: | bromodomain containing 4 |
Gene Aliases: |
Brd5, HUNK1, MCAP, WI-11513 |
Location: |
Chr.17:32196272-32284722 on Build GRCm38 |
Assay Gene Location: | Overlaps - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brd4 | NM_001286630.1 | NP_001273559.1 | ||
| NM_020508.4 | NP_065254.3 | |||
| NM_198094.2 | NP_932762.2 | |||
| AF461395.1 | AAL67833.1 | |||
| AF461396.1 | AAL67834.1 | |||
| AK042431.1 | ||||
| AK147545.1 | BAE27987.1 | |||
| AK154514.1 | BAE32643.1 | |||
| BC082782.1 | AAH82782.1 | |||
| BC103773.1 | AAI03774.1 | |||
| BC138834.1 | ||||
| BC138835.1 | ||||
| BY779550.1 |

Genomic Map