Assay Details
Target Gene Details
Entrez Gene ID: | 78309 |
Gene Name: | cullin 9 |
Gene Aliases: |
1810035I07Rik, Cul-9, Parc, mKIAA0708 |
Location: |
Chr.17:46500609-46546388 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cul9 | NM_001081335.2 | NP_001074804.2 | ||
| AK051474.1 | ||||
| AK122351.1 | BAC65633.1 | |||
| AK147675.1 | ||||
| AK162968.1 | ||||
| BP753198.1 |

Genomic Map