Assay Details
Target Gene Details
Entrez Gene ID: | 76872 |
Gene Name: | coiled-coil domain containing 116 |
Gene Aliases: |
4930432J16Rik |
Location: |
Chr.16:17139064-17147146 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc116 | AK141314.1 | 1 | 550 | |
| BC057359.1 | 1 | 177 | AAH57359.1 |
Target Gene Details
Entrez Gene ID: | 69101 |
Gene Name: | YdjC homolog (bacterial) |
Gene Aliases: |
1810015A11Rik, 4930521M19Rik |
Location: |
Chr.16:17146882-17148857 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ydjc | NM_026940.4 | 1 | 97 | NP_081216.1 |
| AK007503.1 | 1 | 86 | BAB25075.1 | |
| AK141482.1 | 1 | 86 | ||
| BC115584.1 | 1 | 85 | ||
| BC115585.1 | 1 | 85 |

Genomic Map