Assay Details
Target Gene Details
Entrez Gene ID: | 11629 |
Gene Name: | allograft inflammatory factor 1 |
Gene Aliases: |
AI607846, AIF-1, D17H6S50E, G1, Iba1 |
Location: |
Chr.17:35170991-35176087 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aif1 | NM_019467.2 | NP_062340.1 | ||
| AB013745.1 | BAA28216.1 | |||
| AF074959.1 | AAC25604.1 | |||
| AK006184.1 | BAB24445.1 | |||
| AK006562.1 | BAB24654.1 | |||
| BC021539.1 | AAH21539.1 | |||
| CT010265.1 | CAJ18473.1 | |||
| CT010385.1 | CAJ18592.1 | |||
| D86382.1 | BAA86387.1 |
More Information
Set Membership: |
|

Genomic Map