Assay Details
Target Gene Details
Entrez Gene ID: | 12815 |
Gene Name: | collagen, type XI, alpha 2 |
Gene Aliases: |
- |
Location: |
Chr.17:34038486-34066684 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col11a2 | NM_009926.1 | 4 | 744 | NP_034056.1 |
| D38412.1 | 4 | 708 | BAA18910.1 | |
| KJ650220.1 | 4 | 526 | ||
| U16789.1 | 4 | 745 | AAA67751.1 |
More Information
Set Membership: |
|

Genomic Map