Assay Details
Target Gene Details
Entrez Gene ID: | 13079 |
Gene Name: | cytochrome P450, family 21, subfamily a, polypeptide 1 |
Gene Aliases: |
21-OH, 21OH, 21OHA, 21OHB, CYP21OH-A, Cyp21, Cyp21-ps1, Cyp21B, Cyp21a2-ps, Cyp21a2ps, Oh21-1, Oh21-2 |
Location: |
Chr.17:34801348-34804426 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp21a1 | NM_009995.2 | NP_034125.2 | ||
| AK146565.1 | BAE27263.1 | |||
| BC127167.1 |

Genomic Map