Assay Details
Target Gene Details
Entrez Gene ID: | 72775 |
Gene Name: | Fanconi anemia, complementation group E |
Gene Aliases: |
2810451D06Rik, AI415634, AW209126 |
Location: |
Chr.17:28313530-28326574 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fance | NM_001163819.1 | NP_001157291.1 | ||
| NM_001163820.1 | NP_001157292.1 | |||
| NR_028296.1 | ||||
| NR_028297.1 | ||||
| AK013325.1 | ||||
| AK135017.1 | BAE22385.1 | |||
| AK172283.1 | BAE42924.1 | |||
| AK180906.1 | 2 | 276 | ||
| AK187114.1 | ||||
| AK209410.1 | ||||
| AK217065.1 | ||||
| BC016538.1 | AAH16538.1 |
More Information
Set Membership: |
|

Genomic Map