Assay Details
Target Gene Details
Entrez Gene ID: | 74112 |
Gene Name: | ubiquitin specific peptidase 16 |
Gene Aliases: |
1200004E02Rik, 2810483I07Rik, 6330514E22Rik, UBPM |
Location: |
Chr.16:87454708-87483515 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp16 | NM_024258.2 | NP_077220.2 | ||
| AK004594.2 | ||||
| AK013437.1 | ||||
| AK158778.1 | 1 | 1819 | ||
| AK160038.1 | BAE35580.1 | |||
| BC003278.1 | AAH03278.1 |
More Information
Set Membership: |
|

Genomic Map