Assay Details
Target Gene Details
Entrez Gene ID: | 74112 |
Gene Name: | ubiquitin specific peptidase 16 |
Gene Aliases: |
1200004E02Rik, 2810483I07Rik, 6330514E22Rik, UBPM |
Location: |
Chr.16:87454708-87483515 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp16 | NM_024258.2 | 16 | 2309 | NP_077220.2 |
| AK004594.2 | 16 | 2284 | ||
| AK013437.1 | 10 | 1325 | ||
| AK160038.1 | 16 | 2279 | BAE35580.1 | |
| BC003278.1 | 16 | 2238 | AAH03278.1 | |
| BC004577.1 | 4 | 821 | AAH04577.1 |
More Information
Set Membership: |
|

Genomic Map