Assay Details
Target Gene Details
Entrez Gene ID: | 72016 |
Gene Name: | RIKEN cDNA 1600002H07 gene |
Gene Aliases: |
- |
Location: |
Chr.17:24215054-24220938 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1600002H07Rik | NM_028056.1 | 9 | 1142 | NP_082332.1 |
| AK005395.1 | 8 | 1342 | BAB23999.1 | |
| AK146539.1 | 9 | 1136 | BAE27244.1 | |
| BC072612.1 | 9 | 1142 | AAH72612.1 |

Genomic Map