Assay Details
Target Gene Details
Entrez Gene ID: | 67993 |
Gene Name: | nudix (nucleoside diphosphate linked moiety X)-type motif 12 |
Gene Aliases: |
0610016O18Rik |
Location: |
Chr.17:59000107-59013367 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nudt12 | NM_026497.2 | NP_080773.1 | ||
| AK002641.1 | BAB22253.1 | |||
| AK083846.1 | ||||
| AK147867.1 | BAE28190.1 | |||
| BC057657.1 | AAH57657.1 |
More Information
Set Membership: |
|

Genomic Map