Assay Details
Target Gene Details
Entrez Gene ID: | 53414 |
Gene Name: | bystin-like |
Gene Aliases: |
Bys, Enp1 |
Location: |
Chr.17:47599331-47611492 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bysl | NM_016859.3 | NP_058555.3 | ||
| AF007802.1 | AAB94491.1 | |||
| AK019239.1 | BAB31619.2 | |||
| AK032993.1 | BAC28118.1 | |||
| AK049195.1 | BAC33603.1 | |||
| AK051704.1 | BAC34727.1 | |||
| AK083033.1 | BAC38736.1 | |||
| AK143027.1 | BAE25255.1 | |||
| AK168301.1 | BAE40242.1 | |||
| BC017530.1 |

Genomic Map