Assay Details
Target Gene Details
Entrez Gene ID: | 207209 |
Gene Name: | coiled-coil domain containing 154 |
Gene Aliases: |
Ccdc157, Gm317 |
Location: |
Chr.17:25162168-25171913 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc154 | NM_001079929.2 | NP_001073398.2 | ||
| AK133084.1 | BAE21504.1 | |||
| JN935900.1 |

Genomic Map