Assay Details
Target Gene Details
Entrez Gene ID: | 12648 |
Gene Name: | chromodomain helicase DNA binding protein 1 |
Gene Aliases: |
4930525N21Rik, AI851787, AW555109 |
Location: |
Chr.17:15704967-15772612 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chd1 | NM_007690.3 | 16 | 2749 | NP_031716.2 |
| AK015218.1 | 3 | 409 | BAB29753.1 | |
| AK049686.1 | 16 | 2744 | BAC33872.1 | |
| AK140386.1 | 16 | 2729 | BAE24366.1 | |
| BC115822.1 | 15 | 2387 | ||
| L10410.1 | 16 | 2426 | AAB08486.1 |

Genomic Map