Assay Details
Target Gene Details
Entrez Gene ID: | 26378 |
Gene Name: | 2-4-dienoyl-Coenzyme A reductase 2, peroxisomal |
Gene Aliases: |
- |
Location: |
Chr.17:26081208-26091517 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Decr2 | NM_011933.2 | NP_036063.1 | ||
| AF155575.1 | AAD38196.1 | |||
| AK137212.1 | BAE23272.1 | |||
| AK149270.1 | BAE28781.1 | |||
| AK162961.1 | BAE37137.1 | |||
| BC021865.1 | AAH21865.1 |
More Information
Set Membership: |
|

Genomic Map