Assay Details
Target Gene Details
Entrez Gene ID: | 70785 |
Gene Name: | DENN/MADD domain containing 1C |
Gene Aliases: |
4432409M07Rik |
Location: |
Chr.17:57066051-57078510 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dennd1c | NM_153551.1 | NP_705779.1 | ||
| AK040537.1 | ||||
| BC035208.1 | AAH35208.1 |

Genomic Map