Assay Details
Target Gene Details
Entrez Gene ID: | 224742 |
Gene Name: | ATP-binding cassette, sub-family F (GCN20), member 1 |
Gene Aliases: |
AU041969, Abc50, D17Wsu166e, GCN20 |
Location: |
Chr.17:35956816-35969800 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcf1 | NM_013854.1 | NP_038882.1 | ||
| AK145549.1 | ||||
| BC046965.1 | AAH46965.1 | |||
| BC063094.1 | AAH63094.1 |
Target Gene Details
Entrez Gene ID: | 100124459 |
Gene Name: | microRNA 877 |
Gene Aliases: |
Mirn877, mir-877, mmu-mir-877 |
Location: |
Chr.17:35960730-35960814 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mir877 | NR_030608.1 |
More Information
Set Membership: |
|

Genomic Map