Assay Details
Target Gene Details
Entrez Gene ID: | 212733 |
Gene Name: | coiled-coil domain containing 64B |
Gene Aliases: |
BICDR-2 |
Location: |
Chr.17:23660523-23668818 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc64b | NM_153784.2 | 3 | 478 | NP_722479.1 |
| AK036793.1 | 3 | 493 | BAC29578.1 | |
| AK133729.1 | 3 | 495 | BAE21806.1 | |
| BC038613.1 | 3 | 504 | AAH38613.1 |
More Information
Set Membership: |
|

Genomic Map