Assay Details
Target Gene Details
Entrez Gene ID: | 12648 |
Gene Name: | chromodomain helicase DNA binding protein 1 |
Gene Aliases: |
4930525N21Rik, AI851787, AW555109 |
Location: |
Chr.17:15704967-15772612 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chd1 | NM_007690.3 | 6 | 1004 | NP_031716.2 |
| AK021188.1 | 6 | 945 | BAB32322.3 | |
| AK042202.1 | 6 | 1005 | BAC31196.1 | |
| AK049686.1 | 6 | 999 | BAC33872.1 | |
| AK140386.1 | 6 | 984 | BAE24366.1 | |
| AK142471.1 | 6 | 954 | BAE25077.1 | |
| BC115822.1 | 5 | 642 | ||
| L10410.1 | 6 | 681 | AAB08486.1 |

Genomic Map