Assay Details
Target Gene Details
Entrez Gene ID: | 106648 |
Gene Name: | cytochrome P450, family 4, subfamily f, polypeptide 15 |
Gene Aliases: |
AI787289, Cyp4f-15 |
Location: |
Chr.17:32685601-32703352 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4f15 | NM_134127.1 | 2 | 249 | NP_598888.1 |
| AF233645.1 | 2 | 249 | AAK15011.1 | |
| BC021377.1 | 2 | 240 | AAH21377.1 |
More Information
Set Membership: |
|

Genomic Map