Assay Details
Target Gene Details
Entrez Gene ID: | 70101 |
Gene Name: | cytochrome P450, family 4, subfamily f, polypeptide 16 |
Gene Aliases: |
2310021J05Rik, AI746515 |
Location: |
Chr.17:32536545-32551798 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4f16 | NM_024442.1 | NP_077762.1 | ||
| AF233646.1 | AAK15012.1 | |||
| AK078557.1 | BAC37334.1 | |||
| AK083886.1 | ||||
| AK146232.1 | BAE26998.1 | |||
| BC026539.1 | AAH26539.1 |
More Information
Set Membership: |
|

Genomic Map