Assay Details
Target Gene Details
Entrez Gene ID: | 245847 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
5730457F11Rik |
Location: |
Chr.17:24155833-24163733 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amdhd2 | NM_172935.4 | 5 | 554 | NP_766523.2 |
| AK077584.1 | 5 | 537 | BAC36877.1 | |
| AK090009.1 | 5 | 556 | BAC41042.1 | |
| AK154209.1 | 5 | 554 | ||
| AK159860.1 | 5 | 551 | BAE35434.1 | |
| AK168432.1 | 5 | 512 | BAE40339.1 | |
| AK201857.1 | ||||
| BC037005.1 | 5 | 590 | AAH37005.1 |
More Information
Set Membership: |
|

Genomic Map