Assay Details
Target Gene Details
Entrez Gene ID: | 106585 |
Gene Name: | ankyrin repeat domain 12 |
Gene Aliases: |
2900001A12Rik, AI447928, ANCO-2, AV347965, GAC-1, mKIAA0874 |
Location: |
Chr.17:65965703-66077046 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd12 | NM_001025572.1 | 10 | 5864 | NP_001020743.1 |
| AK035216.1 | 2 | 246 | BAC28981.1 | |
| AK045925.1 | 2 | 899 | BAC32535.1 | |
| AK122392.1 | 2 | 2225 | BAC65674.1 |

Genomic Map