Assay Details
Target Gene Details
Entrez Gene ID: | 208285 |
Gene Name: | cytochrome P450, family 4, subfamily f, polypeptide 17 |
Gene Aliases: |
EG208285 |
Location: |
Chr.17:32506424-32529714 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4f17 | NM_001101445.1 | NP_001094915.1 |
More Information
Set Membership: |
|

Genomic Map