Assay Details
Target Gene Details
Entrez Gene ID: | 106585 |
Gene Name: | ankyrin repeat domain 12 |
Gene Aliases: |
2900001A12Rik, AI447928, ANCO-2, AV347965, GAC-1, mKIAA0874 |
Location: |
Chr.17:65965703-66077046 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd12 | NM_001025572.1 | NP_001020743.1 | ||
| AK012645.1 | 4 | 605 | BAB28377.1 | |
| AK013457.1 | 4 | 656 | BAB28865.1 | |
| AK133715.1 | BAE21795.1 | |||
| AK153850.1 | BAE32211.1 | |||
| BC023046.1 | ||||
| BC050185.1 | AAH50185.1 | |||
| BC064777.1 | AAH64777.1 | |||
| BC080825.1 | AAH80825.1 | |||
| BC094292.1 | 4 | 602 | AAH94292.1 |

Genomic Map