Assay Details
Target Gene Details
Entrez Gene ID: | 320997 |
Gene Name: | cytochrome P450, family 4, subfamily f, polypeptide 39 |
Gene Aliases: |
4732474A20Rik |
Location: |
Chr.17:32452695-32493320 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4f39 | NM_177307.3 | NP_796281.1 | ||
| AK028950.1 | BAC26210.1 | |||
| AK037192.1 | BAC29745.1 | |||
| BC145755.1 |
More Information
Set Membership: |
|

Genomic Map