Assay Details
Target Gene Details
Entrez Gene ID: | 224650 |
Gene Name: | ankyrin repeat and SAM domain containing 1 |
Gene Aliases: |
Anks1a, mKIAA0229 |
Location: |
Chr.17:27909306-28062779 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anks1 | NM_001286040.1 | NP_001272969.1 | ||
| NM_001286041.1 | NP_001272970.1 | |||
| NM_181413.4 | NP_852078.1 | |||
| AK129094.1 | BAC97904.1 | |||
| AK147273.1 | BAE27811.1 | |||
| BC050847.1 | AAH50847.1 |
More Information
Set Membership: |
|

Genomic Map