Assay Details
Target Gene Details
Entrez Gene ID: | 631304 |
Gene Name: | cytochrome P450, family 4, subfamily f, polypeptide 40 |
Gene Aliases: |
EG631304 |
Location: |
Chr.17:32658904-32676919 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp4f40 | NM_001101588.1 | NP_001095058.1 | ||
| BC158089.1 |
More Information
Set Membership: |
|

Genomic Map