Assay Details
Target Gene Details
Entrez Gene ID: | 13819 |
Gene Name: | endothelial PAS domain protein 1 |
Gene Aliases: |
HIF-2alpha, HIF2A, HLF, HRF, MOP2, bHLHe73 |
Location: |
Chr.17:86753864-86833410 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epas1 | NM_010137.3 | 13 | 2533 | NP_034267.3 |
| AF045160.1 | 13 | 2297 | AAC12871.1 | |
| AK136911.1 | 13 | 2537 | ||
| AK141185.1 | 13 | 2268 | ||
| AK158430.1 | 13 | 2697 | BAE34506.1 | |
| BC057870.1 | 13 | 2255 | AAH57870.1 | |
| D89787.1 | 13 | 2495 | BAA20130.1 | |
| U81983.1 | 13 | 2216 | AAB41496.1 |
More Information
Set Membership: |
|

Genomic Map