Assay Details
Target Gene Details
Entrez Gene ID: | 268970 |
Gene Name: | Rho GTPase activating protein 28 |
Gene Aliases: |
AU044757, AW550892, E130310N06 |
Location: |
Chr.17:67842706-68004108 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap28 | NM_172964.4 | 13 | 1698 | NP_766552.3 |
| AK087523.1 | 13 | 1711 | BAC39911.1 | |
| AK133720.1 | 13 | 1537 | BAE21798.1 | |
| AK141901.1 | 13 | 1648 | BAE24877.1 | |
| AK162051.1 | 7 | 738 | BAE36698.1 | |
| AK173147.1 | 12 | 1325 | BAD32425.1 | |
| BC066788.1 | 13 | 1688 | AAH66788.1 |
More Information
Set Membership: |
|

Genomic Map